Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012182.2 | NP_036314.2 | 2 | 1005 | ||
GenBank mRNA | AF071554.1 | - | 1 | 550 | ||
AK290944.1 | - | 2 | 1005 | |||
AK314075.1 | - | 2 | 833 | |||
BC113710.1 | - | 1 | 588 | 8322762 |