Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSGIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182981.2 | NP_892026.1 | 3 | 299 | ||
GenBank mRNA | AK290383.1 | - | 3 | 281 | ||
AY037158.1 | - | 3 | 264 | |||
AY258066.1 | - | 4 | 750 | |||
AY258067.1 | - | 3 | 590 | |||
AY258068.1 | - | 3 | 280 | |||
BC020434.1 | - | 3 | 265 | |||
BC032476.1 | - | 3 | 264 | 5215262 | ||
BC093687.1 | - | 3 | 182 | 7939532 | ||
BC113417.1 | - | 3 | 182 | 8322469 |