Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ012833.1 | - | 1, 2 | 236 | ||
AJ012834.1 | - | 1, 2 | 320 | |||
AJ012835.1 | - | 1, 2 | 322 | |||
AJ223040.1 | - | 1, 2 | 331 | |||
AJ223041.1 | - | 1, 2 | 319 | |||
AY679088.1 | - | 1, 2 | 319 |