Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020994.4 | NP_066274.2 | 2 | 948 | ||
NM_172377.4 | NP_758965.2 | 3 | 719 | |||
GenBank mRNA | AJ012833.1 | - | 3 | 624 | ||
AJ012834.1 | - | 3 | 708 | |||
AJ012835.1 | - | 2 | 939 | |||
AJ223040.1 | - | 2 | 948 | |||
AJ223041.1 | - | 3 | 707 | |||
AY679088.1 | - | 3 | 707 | |||
AY679089.1 | - | 3 | 707 | |||
BC002833.2 | - | 2 | 902 | 3638129 |
Gene Symbol: CTAG1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139250.1 | NP_640343.1 | 3 | 708 |
Gene Symbol: CTAG1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001327.2 | NP_001318.1 | 3 | 708 | ||
GenBank mRNA | AF038567.1 | - | 3 | 743 | ||
AJ003149.1 | - | 3 | 705 | |||
AJ275978.1 | - | 2 | 816 | |||
U87459.1 | - | 3 | 708 |