Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005490.2 | NP_005481.2 | 5 | 859 | ||
XM_005259467.1 | XP_005259524.1 | 5 | 809 | |||
XM_005259472.2 | XP_005259529.1 | 4 | 422 | |||
XM_011527608.2 | XP_011525910.1 | 4 | 1226 | |||
XM_011527609.2 | XP_011525911.1 | 4 | 723 | |||
XM_017026123.1 | XP_016881612.1 | 5 | 809 | |||
GenBank mRNA | AF124249.1 | - | 5 | 816 | ||
AK225386.1 | - | 5 | 782 | |||
AK290293.1 | - | 5 | 935 | |||
AK292781.1 | - | 5 | 782 | |||
AK299406.1 | - | 4 | 432 | |||
AK310659.1 | - | 5 | 782 | |||
AY358406.1 | - | 5 | 816 | |||
BC006281.2 | - | 5 | 796 | 3958342 |