Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015964.3 | NP_057048.2 | 3, 4 | 503 | ||
NM_016140.3 | NP_057224.2 | 4, 5 | 594 | |||
GenBank mRNA | AF078846.1 | - | 3, 4 | 434 | ||
AF132972.1 | - | 3, 4 | 423 | |||
BC000691.2 | - | 3, 4 | 450 | 3349849 | ||
BI754230.1 | - | 4, 5 | 577 |