Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018066.3 | NP_060536.3 | 4 | 1092 | ||
GenBank mRNA | AK001211.1 | - | 4 | 1011 | ||
AK022535.1 | - | 4 | 997 | |||
AY358864.1 | - | 4 | 1001 | |||
AY364261.1 | - | 4 | 810 | |||
BC007815.2 | - | 4 | 934 | 4299794 | ||
BC008634.1 | - | 4 | 958 | 3463813 | ||
BC020272.1 | - | 4 | 977 |