Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FBLIM1 SIRNA Assays ›
Gene Symbol:
FBLIM1
Gene Name:
filamin binding LIM protein 1
Gene Aliases:
CAL, FBLP-1, FBLP1
Chromosome Location:
Chr.1: 15756171 - 15786590 on Build GRCh38
UniGene ID:
Hs.530101
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s195249
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FBLIM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001024215.1
NP_001019386.1
6
1070
NM_001024216.1
NP_001019387.1
3
592
NM_017556.2
NP_060026.2
7
1162
XM_005245900.1
XP_005245957.1
7
1164
XM_005245901.1
XP_005245958.1
7
1130
XM_005245902.1
XP_005245959.1
8
1352
XM_005245903.1
XP_005245960.1
8
1373
XM_005245909.3
XP_005245966.1
8
1474
XM_006710704.3
XP_006710767.1
8
1792
XM_006710705.1
XP_006710768.1
8
1384
XM_011541616.2
XP_011539918.1
8
1789
XM_011541617.2
XP_011539919.1
8
1373
XM_011541618.2
XP_011539920.1
8
1373
XM_017001519.1
XP_016857008.1
8
1679
XM_017001520.1
XP_016857009.1
8
1656
XM_017001521.1
XP_016857010.1
8
1682
XM_017001522.1
XP_016857011.1
8
1653
XM_017001523.1
XP_016857012.1
7
1029
XM_017001524.1
XP_016857013.1
7
1000
XM_017001525.1
XP_016857014.1
8
1396
XM_017001526.1
XP_016857015.1
8
1373
GenBank mRNA
AF459643.1
-
6
1070
AK027444.1
-
3
610
AK055259.1
-
7
1503
AL133035.1
-
3
345
AY180161.1
-
5
863
BC019895.1
-
8
1306
5139869
CN278745.1
-
2
243
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase modulator
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell shape
single organismal cell-cell adhesion
regulation of integrin activation
cell junction assembly
protein binding
zinc ion binding
filamin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0