Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017854.1 | NP_060324.1 | 2 | 278 | ||
XM_017026917.1 | XP_016882406.1 | 2 | 573 | |||
GenBank mRNA | AK000519.1 | - | 2 | 278 | ||
BC002907.2 | - | 2 | 305 | 3941377 |