Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HES6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142853.2 | NP_001136325.1 | 4 | 1408 | ||
NM_001282434.1 | NP_001269363.1 | 4 | 1374 | |||
NM_018645.5 | NP_061115.2 | 4 | 1414 | |||
GenBank mRNA | AF260237.1 | - | 4 | 1255 | ||
AK075040.1 | - | 3 | 1563 | |||
BC007939.2 | - | 4 | 1319 | 4131881 |