Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017397.1 | NP_001017397.1 | 1 | 560 | ||
NM_001017398.1 | NP_001017398.1 | 1 | 560 | |||
NM_001300752.1 | NP_001287681.1 | 1 | 560 | |||
NM_018700.3 | NP_061170.2 | 1 | 560 | |||
XM_017009621.1 | XP_016865110.1 | 1 | 521 | |||
XM_017009623.1 | XP_016865112.1 | 1 | 521 | |||
GenBank mRNA | AB196558.1 | - | 1 | 51 | ||
AI142389.1 | - | 1 | 317 | |||
AJ272269.1 | - | 1 | 177 | |||
AK125426.1 | - | 1 | 337 | |||
BC045164.1 | - | 1 | 362 | |||
BC046096.1 | - | 1 | 508 | |||
BC130334.1 | - | 1 | 229 | 40146365 |