Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005493.2 | NP_005484.2 | 10 | 1661 | ||
XM_006714945.2 | XP_006715008.1 | 10 | 1141 | |||
XM_017010149.1 | XP_016865638.1 | 10 | 1049 | |||
GenBank mRNA | AB008515.1 | - | 10 | 1350 | ||
AB055311.1 | - | 10 | 1661 | |||
AF306510.1 | - | 10 | 1199 | |||
AK313334.1 | - | 9 | 1052 | |||
BC019886.1 | - | 10 | 1115 | |||
BC052781.1 | - | 10 | 1822 | |||
BC063849.1 | - | 8 | 917 | 6186252 | ||
KU921435.1 | - | 5 | 675 |