Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB038783.1 | - | 9 | 2471 |
Gene Symbol: MUC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005960.1 | NP_005951.1 | 10 | 10011 | ||
XM_011516246.2 | XP_011514548.1 | 11 | 10340 | |||
XM_011516247.2 | XP_011514549.1 | 11 | 10290 | |||
XM_011516249.2 | XP_011514551.1 | 11 | 10141 | |||
XM_011516250.2 | XP_011514552.1 | 11 | 9629 | |||
XM_017012231.1 | XP_016867720.1 | 11 | 9837 | |||
GenBank mRNA | AB038784.1 | - | 9 | 2468 | ||
AF007194.1 | - | 10 | 3370 | |||
AF143371.1 | - | 8 | 935 | |||
AF143372.1 | - | 7 | 761 | |||
AF143373.1 | - | 7 | 819 |