Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022358.3 | NP_071753.2 | 2 | 1104 | ||
GenBank mRNA | AF257081.1 | - | 2 | 1104 | ||
AF294350.1 | - | 2 | 1040 | |||
AF294351.1 | - | 2 | 1040 | |||
BC093874.1 | - | 2 | 1084 | 7939719 | ||
BC143282.1 | - | 2 | 1084 | 9051785 |