Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VKORC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311311.1 | NP_001298240.1 | 4 | 772 | ||
NM_024006.5 | NP_076869.1 | 3 | 688 | |||
NM_206824.2 | NP_996560.1 | 2 | 578 | |||
GenBank mRNA | AF176924.1 | - | 4 | 712 | ||
AK129513.1 | - | 2 | 376 | |||
AK289790.1 | - | 2 | 477 | |||
AK312005.1 | - | 3 | 601 | |||
AY423044.1 | - | 3 | 418 | |||
AY466113.1 | - | 3 | 630 | |||
AY521634.1 | - | 3 | 418 | |||
BC000828.1 | - | 3 | 418 | |||
BC002911.2 | - | 3 | 456 | 3944530 | ||
BI822140.1 | - | 3 | 644 | |||
CD249837.1 | - | 379 | ||||
CN484751.1 | - | 4 | 499 | |||
KU178762.1 | - | 3 | 418 | |||
KU178763.1 | - | 2 | 308 |