Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002800.2 | NP_001002800.1 | 11 | 1989 | ||
NM_001288753.1 | NP_001275682.1 | 11 | 1914 | |||
NM_005496.3 | NP_005487.3 | 10 | 1806 | |||
XM_011512311.2 | XP_011510613.1 | 11 | 1877 | |||
XM_011512312.2 | XP_011510614.1 | 10 | 1498 | |||
XM_017005491.1 | XP_016860980.1 | 9 | 1720 | |||
GenBank mRNA | AB019987.1 | - | 11 | 1650 | ||
AF092564.1 | - | 9 | 1280 | |||
AK122939.1 | - | 3 | 762 | |||
AK128143.1 | - | 11 | 1568 | |||
AK307508.1 | - | 1105 | ||||
AK315070.1 | - | 11 | 1645 | |||
AL136877.1 | - | 10 | 1770 | |||
AL833949.1 | - | 10 | 2697 |