Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199120.1 | NP_001186049.1 | 5 | 522 | ||
NM_001199121.1 | NP_001186050.1 | 5 | 505 | |||
NM_024839.2 | NP_079115.1 | 5 | 498 | |||
GenBank mRNA | AF212152.2 | - | 5 | 468 | ||
AJ504713.1 | - | 5 | 458 | |||
AJ504714.1 | - | 4 | 537 | |||
AJ504715.1 | - | 4 | 527 | |||
AJ504716.1 | - | 5 | 465 | |||
AK026291.1 | - | 5 | 467 | |||
BC011730.2 | - | 5 | 460 | 4422871 | ||
BG708140.1 | - | 5 | 529 | |||
BI598757.1 | - | 5 | 525 | |||
DA270919.1 | - | 5 | 498 |
Gene Symbol: TRIM39-RPP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199119.1 | NP_001186048.1 | 10 | 1509 | ||
GenBank mRNA | EU012025.1 | - | 10 | 1238 |