Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024980.4 | NP_079256.4 | 3 | 810 | ||
XM_005263496.4 | XP_005263553.1 | 3 | 720 | |||
GenBank mRNA | AK292888.1 | - | 3 | 764 | ||
AY768808.1 | - | 3 | 810 |
Set Membership: |
G-protein coupled receptor |