Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAIAP2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025045.5 | NP_079321.3 | 12 | 1599 | ||
XM_005261751.4 | XP_005261808.1 | 13 | 1600 | |||
XM_011530379.2 | XP_011528681.1 | 13 | 1674 | |||
XM_011530380.2 | XP_011528682.1 | 11 | 1502 | |||
XM_011530381.2 | XP_011528683.1 | 12 | 1493 | |||
XM_011530382.2 | XP_011528684.1 | 12 | 1881 | |||
XM_011530383.2 | XP_011528685.1 | 11 | 1819 | |||
XM_011530384.2 | XP_011528686.1 | 12 | 1757 | |||
XM_011530386.2 | XP_011528688.1 | 6 | 1218 | |||
XM_011530387.2 | XP_011528689.1 | 7 | 1003 | |||
XM_011530388.2 | XP_011528690.1 | 6 | 983 | |||
GenBank mRNA | AK026263.1 | - | 3 | 1570 | ||
AY358164.1 | - | 12 | 1599 |