Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025069.2 | NP_079345.1 | 2 | 1878 | ||
GenBank mRNA | AK024361.1 | - | 2 | 1846 | ||
BC032534.1 | - | 2 | 1807 | 5527569 | ||
BC084581.1 | - | 3 | 559 |