Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002800.2 | NP_001002800.1 | 14 | 2491 | ||
NM_001288753.1 | NP_001275682.1 | 14 | 2416 | |||
NM_005496.3 | NP_005487.3 | 13 | 2308 | |||
XM_006713459.3 | XP_006713522.1 | 13 | 2027 | |||
XM_011512311.2 | XP_011510613.1 | 14 | 2379 | |||
XM_011512312.2 | XP_011510614.1 | 13 | 2000 | |||
XM_017005491.1 | XP_016860980.1 | 12 | 2222 | |||
GenBank mRNA | AB019987.1 | - | 14 | 2152 | ||
AF092564.1 | - | 12 | 1782 | |||
AK122939.1 | - | 6 | 1264 | |||
AK128143.1 | - | 14 | 2070 | |||
AK307508.1 | - | 1607 | ||||
AK315070.1 | - | 14 | 2147 | |||
AL136877.1 | - | 13 | 2272 | |||
AL833949.1 | - | 13 | 3199 |