Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190467.1 | NP_001177396.1 | 6 | 4324 | ||
NR_104345.1 | 7 | 1667 | ||||
GenBank mRNA | AK025837.1 | - | 3 | 2060 | ||
AK096643.1 | - | 3 | 2336 | |||
AL442075.1 | - | 2 | 839 |