Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTYH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025250.2 | NP_079526.1 | 3 | 494 | ||
XM_011515535.2 | XP_011513837.1 | 3 | 462 | |||
XM_017012655.1 | XP_016868144.1 | 3 | 464 | |||
XM_017012656.1 | XP_016868145.1 | 3 | 462 | |||
GenBank mRNA | AB051478.2 | - | 3 | 489 | ||
AB162931.1 | - | 3 | 319 | |||
AL522452.3 | - | 3 | 492 | |||
BC131824.1 | - | 3 | 365 | 40128913 | ||
BC152447.1 | - | 3 | 489 | 8862578 |