Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030943.3 | NP_112205.2 | 5 | 455 | ||
XM_011537202.2 | XP_011535504.1 | 5 | 700 | |||
XM_011537203.2 | XP_011535505.1 | 5 | 956 | |||
GenBank mRNA | AF328788.1 | - | 5 | 444 | ||
AK055802.1 | - | 4 | 1168 | |||
AY358468.1 | - | 5 | 442 | |||
BC029948.1 | - | 5 | 455 |