Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317238.1 | NP_001304167.1 | 10, 11 | 1517 | ||
NM_031921.5 | NP_114127.3 | 12, 13 | 1400 | |||
XM_005244806.3 | XP_005244863.1 | 12, 13 | 1423 | |||
XM_011542241.2 | XP_011540543.1 | 12, 13 | 1423 | |||
XM_011542243.2 | XP_011540545.1 | 11, 12 | 1136 | |||
XM_011542244.1 | XP_011540546.1 | 12, 13 | 1567 | |||
GenBank mRNA | AB033099.1 | - | 10, 11 | 1606 | ||
AF343078.1 | - | 12, 13 | 1333 | |||
AK024028.1 | - | 10, 11 | 985 | |||
AK127857.1 | - | 5, 6 | 2051 | |||
AK290586.1 | - | 12, 13 | 1392 | |||
BC002542.2 | - | 12, 13 | 1326 | 3138578 | ||
BC018701.1 | - | 12, 13 | 1326 | 3139886 |