Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290212.1 | NP_001277141.1 | 4 | 1792 | ||
NM_031945.4 | NP_114151.3 | 3 | 1558 | |||
NR_110866.1 | 4 | 1826 | ||||
GenBank mRNA | AB593081.1 | - | 4 | 1765 | ||
AB593082.1 | - | 3 | 1553 | |||
AF325213.1 | - | 3 | 1514 | |||
BC021923.1 | - | 4 | 1799 | 4876887 | ||
BC032802.1 | - | 3 | 1529 | 5191718 |