Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI27L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032036.2 | NP_114425.1 | 4 | 361 | ||
GenBank mRNA | AF208232.1 | - | 4 | 327 | ||
AF238978.1 | - | 2 | 194 | |||
BC022800.2 | - | 2 | 269 | |||
BC032626.1 | - | 4 | 320 | 5229498 | ||
BE737110.1 | - | 362 |