Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEH2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032193.3 | NP_115569.2 | 4 | 715 | ||
GenBank mRNA | AF346606.1 | - | 4 | 715 | ||
AK024627.1 | - | 4 | 572 | |||
AK225131.1 | - | 4 | 572 | |||
AK297660.1 | - | 4 | 485 | |||
BC023588.2 | - | 4 | 595 | 4763204 | ||
BM921525.1 | - | 4 | 584 | |||
L08647.1 | - | 3 | 444 |