Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1M2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300887.1 | NP_001287816.1 | 9 | 1028 | ||
NM_005498.4 | NP_005489.2 | 9 | 1022 | |||
GenBank mRNA | AF020797.2 | - | 9 | 985 | ||
AK023863.1 | - | 9 | 1023 | |||
AK222946.1 | - | 9 | 988 | |||
AK315689.1 | - | 9 | 1013 | |||
BC003387.2 | - | 9 | 960 | 3446133 | ||
BC003612.2 | - | 9 | 959 | 2988458 | ||
BC005021.1 | - | 9 | 1028 | 3139433 |