Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304353.1 | NP_001291282.1 | 4 | 405 | ||
NM_001304354.1 | NP_001291283.1 | 4 | 402 | |||
NM_032309.3 | NP_115685.1 | 4 | 568 | |||
GenBank mRNA | AK096990.1 | - | 3 | 2911 | ||
BC004498.2 | - | 4 | 546 | 3831856 | ||
BM563093.1 | - | 4 | 402 | |||
BX115079.1 | - | 4 | 404 | |||
N39730.1 | - | 4 | 402 |