Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040160.2 | NP_001035250.1 | 5 | 609 | ||
NM_001040161.2 | NP_001035251.1 | 3 | 386 | |||
NM_001040162.2 | NP_001035252.1 | 4 | 446 | |||
NM_001040165.2 | NP_001035255.1 | 4 | 549 | |||
NM_001288710.1 | NP_001275639.1 | 5 | 617 | |||
NM_032366.4 | NP_115742.3 | 6 | 677 | |||
NR_109976.1 | 5 | 621 | ||||
NR_109977.1 | 5 | 514 | ||||
NR_109978.1 | 6 | 784 | ||||
NR_109979.1 | 4 | 454 | ||||
XM_011522713.2 | XP_011521015.1 | 6 | 876 | |||
XM_011522714.2 | XP_011521016.1 | 5 | 701 | |||
GenBank mRNA | AI146806.1 | - | 4 | 443 | ||
AK057077.1 | - | 3 | 1575 | |||
AK294482.1 | - | 6 | 613 | |||
AK295628.1 | - | 5 | 545 | |||
BC007207.1 | - | 3 | 368 | 4054799 | ||
BI762987.1 | - | 6 | 653 | |||
BM827705.1 | - | 4 | 381 | |||
BP273946.1 | - | 5 | 553 | |||
BQ921757.1 | - | 582 | ||||
BU539308.1 | - | 4 | 402 |