Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288587.2 | NP_001275516.1 | 1 | 590 | ||
NM_001321630.1 | NP_001308559.1 | 1 | 590 | |||
NM_032899.6 | NP_116288.2 | 2 | 1014 | |||
NM_207006.2 | NP_996889.1 | 2 | 2099 | |||
XM_005251087.2 | XP_005251144.1 | 1 | 387 | |||
XM_011517338.1 | XP_011515640.1 | 1 | 387 | |||
GenBank mRNA | AF497803.1 | - | 1 | 339 | ||
AK095301.1 | - | 2 | 2099 | |||
BC007828.2 | - | 1 | 261 | 3610547 | ||
BC052300.1 | - | 1 | 403 | 6421272 | ||
DA454025.1 | - | 1 | 553 | |||
DQ280322.1 | - | 1 | 300 | |||
DQ280323.1 | - | 2 | 987 |