Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAGE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182482.2 | NP_872288.2 | 2, 3 | 328 | ||
GenBank mRNA | AF218570.1 | - | 2, 3 | 328 | ||
AK292540.1 | - | 2, 3 | 368 | |||
BC101132.2 | - | 2, 3 | 223 | |||
BC101133.2 | - | 2, 3 | 223 |
Gene Symbol: BAGE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182481.1 | NP_872287.1 | 2, 3 | 328 | ||
GenBank mRNA | AF339514.1 | - | 2, 3 | 328 | ||
BC101134.2 | - | 2, 3 | 223 | 40014666 |