Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033131.3 | NP_149122.1 | 4 | 2000 | ||
GenBank mRNA | AK056278.1 | - | 4 | 1971 | ||
BC103921.1 | - | 5 | 1200 | 40007188 | ||
BC103923.1 | - | 5 | 1200 | 40007191 |