Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53I13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138349.2 | NP_612358.3 | 7 | 1431 | ||
XM_017025282.1 | XP_016880771.1 | 7 | 1803 | |||
GenBank mRNA | AB110209.1 | - | 7 | 1498 | ||
AK075341.1 | - | 7 | 1462 | |||
AK093030.1 | - | 1 | 2447 | |||
AL137404.1 | - | 1 | 806 | |||
BC001593.2 | - | 7 | 1431 | 2988166 | ||
BC004507.2 | - | 2 | 803 |