Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GADD45GIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052850.3 | NP_443082.2 | 2 | 439 | ||
GenBank mRNA | AF412816.1 | - | 2 | 396 | ||
AF475095.1 | - | 2 | 397 | |||
AF479749.1 | - | 2 | 421 | |||
AJ295986.1 | - | 2 | 380 | |||
AJ429498.1 | - | 2 | 397 | |||
BC004944.1 | - | 2 | 385 | |||
BC013027.2 | - | 2 | 394 | |||
BC013039.2 | - | 2 | 394 | |||
BC069200.1 | - | 2 | 399 | 5467620 | ||
BI459923.1 | - | 2 | 444 |