Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145023.1 | NP_001138495.1 | 6, 7 | 1072 | ||
NM_138355.3 | NP_612364.2 | 6, 7 | 1072 | |||
XM_005257776.4 | XP_005257833.1 | 6, 7 | 1052 | |||
XM_017025286.1 | XP_016880775.1 | 6, 7 | 1000 | |||
GenBank mRNA | AK290646.1 | - | 6, 7 | 975 | ||
AK303835.1 | - | 6, 7 | 1028 | |||
BC002980.1 | - | 6, 7 | 1015 | |||
BC010408.2 | - | 6, 7 | 986 | 3945715 | ||
BC017317.2 | - | 6, 7 | 985 | 3832079 |