Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGTLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178311.2 | NP_842563.1 | 2 | 296 | ||
NM_178312.2 | NP_842564.1 | 2 | 224 | |||
XM_005260863.4 | XP_005260920.1 | 2 | 651 | |||
XM_005260864.4 | XP_005260921.1 | 3 | 804 | |||
XM_005260865.4 | XP_005260922.1 | 3 | 804 | |||
XM_011529391.2 | XP_011527693.1 | 2 | 218 | |||
XM_011529392.2 | XP_011527694.1 | 3 | 743 | |||
XM_011529393.2 | XP_011527695.1 | 2 | 590 | |||
XM_017028126.1 | XP_016883615.1 | 2 | 213 | |||
GenBank mRNA | BC040904.1 | - | 2 | 296 | 5744021 | |
L20491.1 | - | 2 | 224 | |||
L20492.1 | - | 1 | 274 |