Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138431.2 | NP_612440.1 | 5 | 1552 | ||
NR_130120.1 | 4 | 1404 | ||||
XM_011516806.2 | XP_011515108.1 | 5 | 2499 | |||
XM_017013005.1 | XP_016868494.1 | 4 | 2594 | |||
GenBank mRNA | BC011982.1 | - | 5 | 1419 | 3860434 | |
DB520468.1 | - | 3 | 406 |