Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DTX2 SIRNA Assays ›
Gene Symbol:
DTX2
Gene Name:
deltex E3 ubiquitin ligase 2
Gene Aliases:
RNF58
Chromosome Location:
Chr.7: 76461655 - 76505995 on Build GRCh38
UniGene ID:
Hs.187058
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s195603
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DTX2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001102594.1
NP_001096064.1
4
1172
NM_001102595.1
NP_001096065.1
3
1007
NM_001102596.1
NP_001096066.1
2
736
NM_020892.2
NP_065943.2
5
1292
XM_005250131.1
XP_005250188.1
5
1183
XM_005250132.1
XP_005250189.1
3
926
XM_011515772.1
XP_011514074.1
5
1354
XM_011515773.2
XP_011514075.1
5
1781
XM_011515774.1
XP_011514076.1
5
1107
XM_011515775.1
XP_011514077.1
4
1201
XM_017011723.1
XP_016867212.1
4
1230
XM_017011724.1
XP_016867213.1
4
1661
XM_017011725.1
XP_016867214.1
4
1079
XM_017011726.1
XP_016867215.1
4
1172
XM_017011727.1
XP_016867216.1
3
986
XM_017011728.1
XP_016867217.1
2
825
XM_017011729.1
XP_016867218.1
2
825
XM_017011730.1
XP_016867219.1
2
825
GenBank mRNA
AB040961.1
-
4
1094
AK023924.1
-
3
948
AK293226.1
-
3
761
AY225124.1
-
2
736
AY225125.1
-
2
736
BC008856.2
-
3
927
4107018
BC018555.1
-
5
1246
4341568
BC026059.2
-
3
927
4099111
BC093079.1
-
4
1060
30512572
DQ010329.1
-
5
1246
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
Notch signaling pathway
protein ubiquitination
protein binding
zinc ion binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0