Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053049.3 | NP_444277.2 | 2 | 514 | ||
GenBank mRNA | BC069530.1 | - | 1 | 286 | 7262181 | |
BC100867.2 | - | 2 | 431 | 40002991 | ||
BC100868.1 | - | 2 | 431 | 40002995 | ||
BC100869.1 | - | 2 | 431 | 40002997 | ||
BC100870.1 | - | 2 | 431 | 40002998 | ||
BQ776877.1 | - | 2 | 507 | |||
BQ777111.1 | - | 2 | 519 |