Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152326.2 | NP_689539.1 | 4 | 1570 | ||
XM_005267310.2 | XP_005267367.1 | 3 | 1707 | |||
XM_005267311.3 | XP_005267368.1 | 3 | 1339 | |||
GenBank mRNA | AK291868.1 | - | 3 | 1236 | ||
BC015422.1 | - | 2 | 1369 | 4397794 | ||
BC027479.1 | - | 2 | 1087 | 5114925 | ||
BX161460.1 | - | 4 | 1570 |