Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308240.1 | NP_001295169.1 | 3 | 781 | ||
NM_205767.2 | NP_991330.1 | 2 | 585 | |||
XM_011527675.2 | XP_011525977.1 | 2 | 534 | |||
XM_017026248.1 | XP_016881737.1 | 2 | 526 | |||
GenBank mRNA | AA292888.1 | - | 2 | 352 | ||
AJ707830.1 | - | 2 | 331 | |||
AK091768.1 | - | 1 | 1284 | |||
BC009557.1 | - | 2 | 219 | 3899541 | ||
BC042386.1 | - | 2 | 194 | 5288954 | ||
BC052640.1 | - | 2 | 196 | |||
BI827162.1 | - | 2 | 584 | |||
CR748109.1 | - | 2 | 549 |