Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JOSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270639.1 | NP_001257568.1 | 5 | 971 | ||
NM_001270640.1 | NP_001257569.1 | 5 | 968 | |||
NM_001270641.1 | NP_001257570.1 | 4 | 845 | |||
NM_001270686.1 | NP_001257615.1 | 5 | 814 | |||
NM_138334.3 | NP_612207.1 | 5 | 817 | |||
XM_011526434.2 | XP_011524736.1 | 5 | 811 | |||
GenBank mRNA | AF247787.1 | - | 4 | 746 | ||
AK131052.1 | - | 4 | 710 | |||
BC051380.1 | - | 5 | 775 | |||
BC062416.1 | - | 5 | 794 | 6503623 |