Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133261.2 | NP_573568.1 | 3, 4 | 626 | ||
XM_005259492.3 | XP_005259549.1 | 3, 4 | 666 | |||
GenBank mRNA | AB073738.1 | - | 3, 4 | 626 | ||
BC117312.1 | - | 3, 4 | 624 | 40125863 | ||
BC117314.1 | - | 3, 4 | 624 | 40125865 |