Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012644.2 | NP_001012662.1 | 6 | 821 | ||
NM_001320478.1 | NP_001307407.1 | 6 | 478 | |||
NM_001320479.1 | NP_001307408.1 | 6 | 478 | |||
NM_080611.4 | NP_542178.2 | 6 | 817 | |||
NM_177991.2 | NP_817130.1 | 6 | 583 | |||
XM_017027654.1 | XP_016883143.1 | 6 | 675 | |||
XM_017027655.1 | XP_016883144.1 | 6 | 466 | |||
XM_017027656.1 | XP_016883145.1 | 6 | 675 | |||
XM_017027657.1 | XP_016883146.1 | 6 | 675 | |||
XM_017027658.1 | XP_016883147.1 | 6 | 615 | |||
XM_017027659.1 | XP_016883148.1 | 6 | 621 | |||
XM_017027660.1 | XP_016883149.1 | 6 | 503 | |||
XM_017027661.1 | XP_016883150.1 | 5 | 612 | |||
XM_017027662.1 | XP_016883151.1 | 6 | 473 | |||
GenBank mRNA | AK091960.1 | - | 5 | 1017 | ||
AK097430.1 | - | 6 | 480 | |||
BC056911.1 | - | 6 | 634 | 6195838 | ||
BI517878.1 | - | 6 | 550 | |||
BM554314.1 | - | 6 | 443 |