Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012644.2 | NP_001012662.1 | 7 | 1067 | ||
NM_001320478.1 | NP_001307407.1 | 7 | 724 | |||
NM_080611.4 | NP_542178.2 | 7 | 1063 | |||
NM_177991.2 | NP_817130.1 | 7 | 829 | |||
GenBank mRNA | AK091960.1 | - | 6 | 1263 | ||
BC056911.1 | - | 7 | 880 | 6195838 | ||
BM554314.1 | - | 7 | 689 |