Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf144 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080825.3 | NP_543015.1 | 1 | 151 | ||
GenBank mRNA | BC032818.1 | - | 1 | 101 | 5172528 | |
BX378327.2 | - | 1 | 151 | |||
DQ080434.1 | - | 1 | 153 |