Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEU4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167599.2 | NP_001161071.1 | 4 | 2322 | ||
NM_001167600.2 | NP_001161072.1 | 4 | 1999 | |||
NM_001167601.2 | NP_001161073.1 | 4 | 2002 | |||
NM_001167602.2 | NP_001161074.1 | 5 | 2500 | |||
NM_080741.3 | NP_542779.2 | 4 | 2319 | |||
GenBank mRNA | AK025617.1 | - | 5 | 2252 | ||
AK091038.1 | - | 5 | 2497 | |||
AK096992.1 | - | 2 | 3112 | |||
AK289421.1 | - | 4 | 1876 | |||
AY203952.1 | - | 4 | 1855 | |||
BC012899.1 | - | 4 | 2273 | 4156395 | ||
BC105974.1 | - | 4 | 1835 | 4944243 |