Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005694.1 | NP_005685.1 | 3 | 323 | ||
GenBank mRNA | AK054900.1 | - | 1 | 1911 | ||
AK095366.1 | - | 2 | 2610 | |||
BC010933.1 | - | 3 | 315 | 4296970 | ||
BC105280.1 | - | 3 | 313 | 6739165 | ||
BC108317.1 | - | 1 | 330 | 6059346 | ||
L77701.1 | - | 3 | 323 |